Learn More
Medchemexpress LLC HY-B0739A 100mg Medchemexpress, Citicoline (sodium) CAS:33818-15-4 Purity:>98%
SDP
Catalog No. 502082654
Encompass_Preferred
Medchemexpress, HY-B0739A 100mg Citicoline (sodium) CAS:33818-15-4 Citicoline sodium salt is an intermediate in the synthesis of phosphatidylcholine which is a component of cell membranes and also exerts neuroprotective effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2654 Supplier Medchemexpress LLC Supplier No. HYB0739A100MG
Medchemexpress, HY-B0739A 100mg Citicoline (sodium) CAS:33818-15-4 Citicoline sodium salt is an intermediate in the synthesis of phosphatidylcholine which is a component of cell membranes and also exerts neuroprotective effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.