Learn More
Cayman Chemical Citicoline (sodium salt), Purity: ≥95%, CAS: 33818-15-4, 100 mg
SDP
Catalog No. 502969211
Encompass_Preferred
An endogenous intermediate in the synthesis of phosphatidylcholine that demonstrates protective effects in cerebral ischemia, traumatic brain injury, and memory disorders when administered exogenously
Catalog No. 50-296-9211 Supplier Cayman Chemical Supplier No. 14629100 MG
An endogenous intermediate in the synthesis of phosphatidylcholine that demonstrates protective effects in cerebral ischemia, traumatic brain injury, and memory disorders when administered exogenously