Learn More
Cayman Chemical CItIcolInsodIum salt 50mg
SDP
Catalog No. 502969436
Encompass_Preferred
An endogenous intermediate in the synthesis of phosphatidylcholine that demonstrates protective effects in cerebral ischemia, traumatic brain injury, and memory disorders when administered exogenously
Catalog No. 50-296-9436 Supplier Cayman Chemical Supplier No. 1462950 MG
An endogenous intermediate in the synthesis of phosphatidylcholine that demonstrates protective effects in cerebral ischemia, traumatic brain injury, and memory disorders when administered exogenously