Learn More
Electron Microscopy Sciences Citifluor™ Mountant Solution AFR3, Glycerol Free, Non Hardening 10 ML
SDP
Catalog No. 501927393
Encompass_Preferred
Citifluor mounting medium for immunofluorescence is designed to reduce the fading of the fluorescence of dyes used for labeling biological specimens. In the technique of fluorescence microscopy, the fluorescence is stimulated by high intensity UV and/or visible light. Absorption of the light populates an excited state of the dye (usually a singlet state) and this leads to fluorescence. However, the excited state of the dye may undergo chemical reaction, which leads to destruction as evidenced by the fading of the fluorescence and consequent loss of the image. PBS solution containing a NEW non-amine, non phenolic antifadent.
Material: Glycerol Free
Type: Non Hardening
Catalog No. 50-192-7393 Supplier Electron Microscopy Sciences Supplier No. 1797310
Citifluor mounting medium for immunofluorescence is designed to reduce the fading of the fluorescence of dyes used for labeling biological specimens. In the technique of fluorescence microscopy, the fluorescence is stimulated by high intensity UV and/or visible light. Absorption of the light populates an excited state of the dye (usually a singlet state) and this leads to fluorescence. However, the excited state of the dye may undergo chemical reaction, which leads to destruction as evidenced by the fading of the fluorescence and consequent loss of the image. PBS solution containing a NEW non-amine, non phenolic antifadent.
Material: Glycerol Free
Type: Non Hardening