Learn More
Ted Pella Inc CITIFLUOR GLYCEROL MEDIUM 25ML
SDP
Catalog No. NC9371014
Encompass
Citifluor, glycerol, PBS and additives,quantity:25ml,application:Immuno Fluorescent Mountant Media
Catalog No. NC9371014 Supplier Ted Pella Inc Supplier No. 19470
Citifluor, glycerol, PBS and additives,quantity:25ml,application:Immuno Fluorescent Mountant Media