Learn More
Electron Microscopy Sciences Citifluor™ Mountant Solution AF100, Glycerol Free, Hardening 5 ML
SDP
Catalog No. 501928881
Encompass_Preferred
Citifluor mounting medium for immunofluorescence is designed to reduce the fading of the fluorescence of dyes used for labeling biological specimens. In the technique of fluorescence microscopy, the fluorescence is stimulated by high intensity UV and/or visible light. Absorption of the light populates an excited state of the dye (usually a singlet state) and this leads to fluorescence. However, the excited state of the dye may undergo chemical reaction, which leads to destruction as evidenced by the fading of the fluorescence and consequent loss of the image. A PBS solution of an amine-based antifadent that if used as an additive to CFPVOH (1:9).
Material: Glycerol Free
Type: Hardening
Catalog No. 50-192-8881 Supplier Electron Microscopy Sciences Supplier No. AF1005
Citifluor mounting medium for immunofluorescence is designed to reduce the fading of the fluorescence of dyes used for labeling biological specimens. In the technique of fluorescence microscopy, the fluorescence is stimulated by high intensity UV and/or visible light. Absorption of the light populates an excited state of the dye (usually a singlet state) and this leads to fluorescence. However, the excited state of the dye may undergo chemical reaction, which leads to destruction as evidenced by the fading of the fluorescence and consequent loss of the image. A PBS solution of an amine-based antifadent that if used as an additive to CFPVOH (1:9).
Material: Glycerol Free
Type: Hardening