Learn More
Biogenex Laboratories CITRA PLUS ANTIGEN RETRIEVAL
SDP
Catalog No. NC9755543
Encompass_Preferred
Antigen Retrieval Citra Plus Solution (10X Concentrated) 500mL Retrieves Most Antigens in Formalin-Fixed Tissues Excellent Results with ER Antigen
Catalog No. NC9755543 Supplier Biogenex Laboratories Supplier No. HK0809K
Antigen Retrieval Citra Plus Solution (10X Concentrated) 500mL Retrieves Most Antigens in Formalin-Fixed Tissues Excellent Results with ER Antigen