Learn More
Medchemexpress LLC Citraconic anhydride | 616-02-4 | 99.8% | C5H4O3 | 10 G
SDP
Catalog No. 5000434922
Encompass_Preferred
Citraconic anhydride (Methylmaleic anhydride) is a derivative of maleic anhydride and a novel antigen retrieval solution. It reversibly blocks protein amino groups, stabilizing specific enzymes and improving their catalytic performance. It reacts with free amino groups on proteins (especially lysine residues), converting positively charged NH3+ into carboxyl groups, thereby disrupting methylene bridge crosslinks caused by Formaldehyde during antigen retrieval. This product is for research use only.
  • Reversibly blocks protein amino groups
  • Stabilizes specific enzymes and improves catalytic performance
  • Disrupts methylene bridge crosslinks caused by formaldehyde during antigen retrieval
  • Functionalizes isotactic polypropylene
  • Responds precisely to pH changes to achieve reversible modification
Catalog No. 50-004-34922 Supplier Medchemexpress LLC Supplier No. HY7803510G
Add to Cart
Add to Cart
Citraconic anhydride (Methylmaleic anhydride) is a derivative of maleic anhydride and a novel antigen retrieval solution. It reversibly blocks protein amino groups, stabilizing specific enzymes and improving their catalytic performance. It reacts with free amino groups on proteins (especially lysine residues), converting positively charged NH3+ into carboxyl groups, thereby disrupting methylene bridge crosslinks caused by Formaldehyde during antigen retrieval. This product is for research use only.
  • Reversibly blocks protein amino groups
  • Stabilizes specific enzymes and improves catalytic performance
  • Disrupts methylene bridge crosslinks caused by formaldehyde during antigen retrieval
  • Functionalizes isotactic polypropylene
  • Responds precisely to pH changes to achieve reversible modification