Learn More
Medchemexpress LLC Citraconic anhydride | 616-02-4 | 99.8% | C5H4O3 | 50 G
SDP
Catalog No. 5000434823
Encompass_Preferred
Citraconic anhydride is a derivative of maleic anhydride and a novel antigen retrieval solution. It reversibly blocks protein amino groups, stabilizing specific enzymes and improving their catalytic performance. This compound reacts with free amino groups on proteins, particularly lysine residues, converting positively charged NH3+ into carboxyl groups to disrupt methylene bridge crosslinks during antigen retrieval. It also functionalizes isotactic polypropylene and responds to pH changes for reversible modification.
  • Antigen retrieval solution
  • Reversible blocking of protein amino groups to stabilize enzymes and enhance catalytic performance
  • Disruption of methylene bridge crosslinks caused by formaldehyde during antigen retrieval
  • Functionalization of isotactic polypropylene
  • Reversible modification through precise pH response
Catalog No. 50-004-34823 Supplier Medchemexpress LLC Supplier No. HY7803550G
Add to Cart
Add to Cart
Citraconic anhydride is a derivative of maleic anhydride and a novel antigen retrieval solution. It reversibly blocks protein amino groups, stabilizing specific enzymes and improving their catalytic performance. This compound reacts with free amino groups on proteins, particularly lysine residues, converting positively charged NH3+ into carboxyl groups to disrupt methylene bridge crosslinks during antigen retrieval. It also functionalizes isotactic polypropylene and responds to pH changes for reversible modification.
  • Antigen retrieval solution
  • Reversible blocking of protein amino groups to stabilize enzymes and enhance catalytic performance
  • Disruption of methylene bridge crosslinks caused by formaldehyde during antigen retrieval
  • Functionalization of isotactic polypropylene
  • Reversible modification through precise pH response