Learn More
Research Products International Corp Citrajet Low Foaming Acid Cleaner, 1 Gallon Bottle
SDP
Catalog No. 501367627
Encompass
Low foaming, phosphate-free, concentrated, liquid cleaner and metal brightener for high performance removal of metal oxides, scales, salts and inorganic residues in labware washers, parts washers, power-spray systems and manual and ultrasonic cleaning. Corrosion inhibited and free rinsing leaving no interfering residues. Ideal for acid washing and rinsing.
Catalog No. 50-136-7627 Supplier Research Products International Corp Supplier No. 114027
Add to Cart
Add to Cart
Low foaming, phosphate-free, concentrated, liquid cleaner and metal brightener for high performance removal of metal oxides, scales, salts and inorganic residues in labware washers, parts washers, power-spray systems and manual and ultrasonic cleaning. Corrosion inhibited and free rinsing leaving no interfering residues. Ideal for acid washing and rinsing.