Learn More
Electron Microscopy Sciences Citrajet Low Foaming Liquid Acid Cleaner 1 GL
SDP
Catalog No. 501898969
Encompass_Preferred
Low foaming phosphate-free, concentrated, liquid cleaner and metal brightener, high performance removal of metal oxides, scale, salts and inorganic residues in lab-ware washers, parts washers, power-spray systems and manual and ultrasonic cleaning. Corrosion inhibited and free rinsing leaving no interfering residues. Ideal for acid washing and rinsing. Dilute 2:100 and it has a pH 2.5.
Catalog No. 50-189-8969 Supplier Electron Microscopy Sciences Supplier No. 6053204
Low foaming phosphate-free, concentrated, liquid cleaner and metal brightener, high performance removal of metal oxides, scale, salts and inorganic residues in lab-ware washers, parts washers, power-spray systems and manual and ultrasonic cleaning. Corrosion inhibited and free rinsing leaving no interfering residues. Ideal for acid washing and rinsing. Dilute 2:100 and it has a pH 2.5.