Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alconox™ Citrajet™ Low-Foaming Liquid Acid Cleaner
GREENER_CHOICE

Catalog No. 16000145 Shop All Alconox Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
55 gal.
Container Type:
Case
Drum
Jerrican
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Container Type
16-000-145 1 gal. Case
16-000-146 5 gal. Jerrican
16-000-149 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 16-000-145 Supplier Alconox™ Supplier No. 2001
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Liquid cleaner and metal brightener for removal of metal oxides, scales, salts and inorganic residues

 

  • Concentrated, biodegradable and phosphate-free
  • Contains blend of organic acids and surfactants
  • Corrosion inhibited and free rinsing leaving no interfering residues
  • Ideal for acid washing and rinsing
  • Dilute 1:100 for use (10mL in 1L water)
  • Use to pass cleaning validation tests for lab accreditation and plant inspection approval
  • USDA authorized for use in federally inspected meat and poultry plants
  • pH 2.5

 

Used to clean:
Laboratory ware, sampling apparatus, dairy equipment optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal forgings, stampings and castings, industrial parts, pipes, tanks, boilers and reactors.

Used to remove:
Scale, metal oxides, metal complexes, trace inorganics, milkstone, soil, grit, buffing compound, grime, grease, fats, oils, particulates, amphoteric peptides, deposits, chemical and solvents.

Surfaces cleaned:
Glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. Can be used on soft metals such as copper, bronze, aluminum, zinc and magnesium if rinsed promptly.

Cleaning method:
Laboratory warewashers, dishwashers, washer-sterilizers, washer-sanitizers, cart washers, spray washers, electrocleaning, ultrasonic cleaners flow through clean-in-place and manual cleaning if proper skin and eye protection is used.

Shelf life: 2 years from date of manufacture

Order Info

3.8L (1 gal.) 1 gal. makes 189L (50 gal.) cleaning solution.

Specifications

Product Type Low-Foaming Liquid Acid Cleaner
Form Liquid
pH 2.5
Appearance Pale Yellow
Composition Phosphate-Free
Container Type Case
Content And Storage Store closed and upright in a cool dry place at 15° to 30°C.
For Use With (Application) Removal of metal oxides, scales, salts and inorganic residues in labware washers, parts washers, power-spray systems
Quantity 4 Pk.
Volume (English) 1 gal.
Volume (Metric) 3.8 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.