Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Citral

Catalog No. C83007500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C83007500ML 500 mL
C830075ML 5 mL
C83007100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C83007500ML Supplier Sigma Aldrich Supplier No. C83007500ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Citral is a mixture of neral (cis isomer of 3,7-dimethyl-2,6-octadienal) and geranial (trans isomer of 3,7-dimethyl-2,6-octadienal). it exhibits antimicrobial activity and pheromonal effects and is generally used for the synthesis of vitamin a and ionones
  • Citral has been used as an anti-tumor agent against breast cancer cell lines (mcf-7 and mda-mb-231)
  • It shows antipyretic effects which can be studied by systemic administration of lipopolysaccharide (lps) to rats
  • It can undergo 1,3-dipolar cycloaddition reaction with trimethylsilylazide to form 1,2,3-triazolines which possess pharmaceutical properties

Specifications

CAS 5392-40-5
Density 0.888 g/mL (at 25°C (literature))
Boiling Point 229°C (lit.)
Molecular Formula C10H16O
Refractive Index n20/D 1.488 (literature)
Linear Formula (CH3)2 C=CHCH2CH2C(CH3)=CHCHO
MDL Number MFCD00006997
Quantity 500 mL
Synonym Lemonal; 3,7-Dimethyl-2,6-octadienal; Geranial and neral mixture
Molecular Weight (g/mol) 152.23
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.