Learn More
Selleck Chemical LLC Citral S5138-25uL
SDP
Catalog No. 503112167
Encompass_Preferred
Citral (Geranialdehyde) is an -unsaturated aldehyde present in the oils of several plants It is an aroma compound used in perfumery for its citrus effect and is also used as a flavor and for fortifying lemon oil
Catalog No. 50-311-2167 Supplier Selleck Chemical LLC Supplier No. S513825UL
Citral (Geranialdehyde) is an -unsaturated aldehyde present in the oils of several plants It is an aroma compound used in perfumery for its citrus effect and is also used as a flavor and for fortifying lemon oil