Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citral, ≥95.0% (cis + trans, GC), MilliporeSigma™ Supelco™

Catalog No. 111014533 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4533 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4533 Supplier MilliporeSigma Supelco Supplier No. 433181ML
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 5392-40-5
Vapor Pressure 0.2 mmHg (200°C)
Vapor Density 5 (vs. air)
Density 0.888 g/mL (at 25°C)
Flash Point 95°C
Physical Form Neat
Linear Formula (CH3)2 C=CHCH2 CH2 C(CH3)=CHCHO
MDL Number MFCD00006997
Boiling Point 229°C
Synonym 3,7-Dimethyl-2,6-octadienal; Geranial and neral mixture
Type Analytical Standard
Quantity 1 mL
Formula Weight 152.23
Percent Purity ≥95.0% (cis + trans, GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.