Learn More
Medchemexpress LLC Citral (Standard) | 5392-40-5 | MFCD00006997 | 100 MG
SDP
Catalog No. 5000419614
Encompass_Preferred
Citral is an orally active monoterpene compound found in lemon grass essential oil. It functions as a natural ALDH1A inhibitor, capable of inducing apoptosis and cell cycle arrest in breast cancer cell lines. Additionally, Citral exhibits analgesic, anti-injurious, and anti-inflammatory properties.
  • Functions as a natural ALDH1A inhibitor
  • Induces apoptosis and cell cycle arrest
  • Demonstrates analgesic effects
  • Possesses anti-injurious properties
  • Exhibits anti-inflammatory effects
  • Relevant for cancer and immunology research
  • Targets aldehyde dehydrogenase
  • Derived from natural plant sources
Catalog No. 50-004-19614 Supplier Medchemexpress LLC Supplier No. HYN7083100MG
Citral is an orally active monoterpene compound found in lemon grass essential oil. It functions as a natural ALDH1A inhibitor, capable of inducing apoptosis and cell cycle arrest in breast cancer cell lines. Additionally, Citral exhibits analgesic, anti-injurious, and anti-inflammatory properties.
  • Functions as a natural ALDH1A inhibitor
  • Induces apoptosis and cell cycle arrest
  • Demonstrates analgesic effects
  • Possesses anti-injurious properties
  • Exhibits anti-inflammatory effects
  • Relevant for cancer and immunology research
  • Targets aldehyde dehydrogenase
  • Derived from natural plant sources