Learn More
Electron Microscopy Sciences EMS CitraMount Mountant 100 ML
SDP
Catalog No. 501900428
Encompass_Preferred
This butyl acetate and acrylic resin mixture is the first mounting medium developed for coverslipping directly from d-limonene based clearants. Superior results are also with xylene. CitraMount medium is recommended for manual coversliping only as it may cause damage to the valves of automated coverslippers. Clear seal has a drying time of less than 24 hours. Refractive index is 1.42 and Viscosity is 90 cP.
Catalog No. 50-190-0428 Supplier Electron Microscopy Sciences Supplier No. 1800501
This butyl acetate and acrylic resin mixture is the first mounting medium developed for coverslipping directly from d-limonene based clearants. Superior results are also with xylene. CitraMount medium is recommended for manual coversliping only as it may cause damage to the valves of automated coverslippers. Clear seal has a drying time of less than 24 hours. Refractive index is 1.42 and Viscosity is 90 cP.