Learn More
Research Products International Corp Citranox Liquid Acid Cleaner and Detergent, 1 Gallon
SDP
Catalog No. 501367626
Encompass
Phosphate free, concentrated cleaner and metal brightener for high performance removal of metal oxides, scale, salts and inorganic residues in manual and ultrasonic cleaning. Corrosion inhibited and free rinsing for reliable results and no interfering residues. Ideal for trace metal analysis.
Catalog No. 50-136-7626 Supplier Research Products International Corp Supplier No. 114021
Phosphate free, concentrated cleaner and metal brightener for high performance removal of metal oxides, scale, salts and inorganic residues in manual and ultrasonic cleaning. Corrosion inhibited and free rinsing for reliable results and no interfering residues. Ideal for trace metal analysis.