Learn More
Sigma Aldrich Fine Chemicals Biosciences CITRANOXR acid detergent b
Catalog No. 502905443
Encompass
for manual or ultrasonic cleaning of metals and hard surfaces. Removes scale milkstone metal oxides fats oils greases solvents and metallic complexes. Biodegradable.
Catalog No. 50-290-5443 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2732364EA
for manual or ultrasonic cleaning of metals and hard surfaces. Removes scale milkstone metal oxides fats oils greases solvents and metallic complexes. Biodegradable.