Learn More
Sigma Aldrich Fine Chemicals Biosciences CITRANOXR acid detergent b1EA
Catalog No. 502897127
Encompass
for manual or ultrasonic cleaning of metals and hard surfaces. Removes scale milkstone metal oxides fats oils greases solvents and metallic complexes. Biodegradable.
Catalog No. 50-289-7127 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2732361EA
for manual or ultrasonic cleaning of metals and hard surfaces. Removes scale milkstone metal oxides fats oils greases solvents and metallic complexes. Biodegradable.