Learn More
Bioworld Citrate Buffer, 0.5M, pH 4.5, 100 mL
Catalog No. 50255776
Encompass_Preferred
Product DescriptionCitrate buffer solution has a 0.5M concentration at a pH of 4.5. Citrate is used as an anticoagulant, a biological buffer, and is often prepared for antigen retrieval of tissue samples. The solution is designed to break protein cross-links, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhanced stain intensity of antibodies. Citrate has as a calcium chelator that forms complexes to disrupt the tendency of blood to clot.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-255-776 Supplier Bioworld Supplier No. 403200551
Product DescriptionCitrate buffer solution has a 0.5M concentration at a pH of 4.5. Citrate is used as an anticoagulant, a biological buffer, and is often prepared for antigen retrieval of tissue samples. The solution is designed to break protein cross-links, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhanced stain intensity of antibodies. Citrate has as a calcium chelator that forms complexes to disrupt the tendency of blood to clot.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.