Learn More
Sigma Aldrich Fine Chemicals Biosciences Citrate Assay Kit sufficie
Catalog No. 502904536
Encompass
Citrate is an intermediate in the citric acid cycle and is involved in fatty acid synthesis. The Citrate Assay Kit provides a simple and automation-ready procedure for measuring citrate concentration. Citrate is converted into pyruvate which is then oxidized with the conversion of the dye into a colored and fluorescent form. The color intensity at 570 nm or fluorescence intensity at lambdaex/em 530/585 nm is directly proportional to the citrate concentration in the sample. Linear detection range 4 to 400 muM citrate for colorimetric assays and 0.5 to 40 muM for fluorimetric assays.
Catalog No. 50-290-4536 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK3331KT
Citrate is an intermediate in the citric acid cycle and is involved in fatty acid synthesis. The Citrate Assay Kit provides a simple and automation-ready procedure for measuring citrate concentration. Citrate is converted into pyruvate which is then oxidized with the conversion of the dye into a colored and fluorescent form. The color intensity at 570 nm or fluorescence intensity at lambdaex/em 530/585 nm is directly proportional to the citrate concentration in the sample. Linear detection range 4 to 400 muM citrate for colorimetric assays and 0.5 to 40 muM for fluorimetric assays.