Learn More
Electron Microscopy Sciences Citrate Buffer, 0.1 M, 500 mL
SDP
Catalog No. 50980221
Encompass_Preferred
Citric acid and Sodium citrate 0.1M, pH range 4.6-6.2, 0-5°C.
Catalog No. 50-980-221 Supplier Electron Microscopy Sciences Supplier No. 11492
Citric acid and Sodium citrate 0.1M, pH range 4.6-6.2, 0-5°C.