Learn More
Electron Microscopy Sciences Citric acid and Sodium citrate 0.1M with a pH range of 4.6-6.2. Ideal for use with fixation, our buffers are prepared fresh for each order and conveniently packaged.
SDP
Catalog No. NC3666163
Encompass_Preferred
Citric acid and Sodium citrate. The buffers are prepared fresh for each order and conveniently packaged.
Catalog No. NC3666163 Supplier Electron Microscopy Sciences Supplier No. 114921L
Citric acid and Sodium citrate. The buffers are prepared fresh for each order and conveniently packaged.