Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citrate Buffer, pH 6.0, 10x, Antigen Retriever, Sigma-Aldrich

Catalog No. 01258997 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-997 1000 mL
01-258-926 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-997 Supplier MilliporeSigma Supelco Supplier No. C99991000ML
Add to Cart
Add to Cart

Occasionally the buffer may contain a yellowish tinge. This is due to preservative and will not affect the function of the buffer.

Store the prepared 1' Citrate buffer, pH 6. 0, Working Solution at 2 to 8°C.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Citrate Buffer
pH 6
Physical Form Liquid
Quantity 1000 mL
Solution Type Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.