Learn More
Biogenex Laboratories CONCENTRATRED ANTIGEN AR 10
SDP
Catalog No. NC0199715
Encompass_Preferred
Antigen Retrieval AR-10 Solution, 10X Concentrated, 100mL, It provides staining of routine, paraffin sections with BioGenex antibodies such as Caldesmon, CD3 (clone PS1), c-myc Protein, PECAM (clone 9G11), GLEPP1
Catalog No. NC0199715 Supplier Biogenex Laboratories Supplier No. HK08720K
Antigen Retrieval AR-10 Solution, 10X Concentrated, 100mL, It provides staining of routine, paraffin sections with BioGenex antibodies such as Caldesmon, CD3 (clone PS1), c-myc Protein, PECAM (clone 9G11), GLEPP1