Learn More
Bioworld Citrate-Buffered Saline (20X), 1 L
Catalog No. 50255756
Encompass_Preferred
Product DescriptionCitrate-buffered saline has a 20X concentration and is composed of 300 mM sodium citrate, 3M sodium chloride, and is at a pH of 7.0. As a conjugate base of a weak acid, citrate can act as a buffering agent or acidity regulator, resisting changes in pH. It is used for nucleic acid hybridization, blot transfer procedures and nucleic acid preparation.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-255-756 Supplier Bioworld Supplier No. 403200291
Product DescriptionCitrate-buffered saline has a 20X concentration and is composed of 300 mM sodium citrate, 3M sodium chloride, and is at a pH of 7.0. As a conjugate base of a weak acid, citrate can act as a buffering agent or acidity regulator, resisting changes in pH. It is used for nucleic acid hybridization, blot transfer procedures and nucleic acid preparation.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.