Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab, Inc. Citrate Label

Catalog No. 22505461
Change view
Click to view available options
Length (English):
0.375 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
22-505-461 0.375 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-505-461 Supplier Marketlab, Inc. Supplier No. 8018
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heme/Coag Label with Blister Packaging.

  • Paper label
  • Permanent adhesive

Specifications

Adhesive Type Peel and Stick
Coating None
Color Blue
Finish Opaque
Format Label
Material Paper
Print Technology None
Product Type Label
Sterility Non-sterile
Length (English) 0.375 in.
Width (English) 1.625 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.