Learn More
MilliporeSigma CITRATE PH6 BUFF RETRIEVAL SOL
Catalog No. 5052547
Encompass
IHC Select Citrate Buffer pH 6.0, 10x. Presentation:Liquid Citrate Buffer containing 0.1% ProClin 300 as a preservative.
Catalog No. 50-525-47 Supplier MilliporeSigma Supplier No. 21545
IHC Select Citrate Buffer pH 6.0, 10x. Presentation:Liquid Citrate Buffer containing 0.1% ProClin 300 as a preservative.