Learn More
Sigma Aldrich Fine Chemicals Biosciences Citrate-phosphate-dextrose
Catalog No. 502905889
Encompass
Citrate-phosphate-dextrose SOLUTIONon (CPD) is an anticoagulant SOLUTIONon. It has 15% less citrate ion over citrate-phosphate-dextrose SOLUTIONon with adenine (ACD) and provides improved isotonicity oxygen release function and posttransfusion viability for red blood cells.
Catalog No. 50-290-5889 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C716510X50ML
Citrate-phosphate-dextrose SOLUTIONon (CPD) is an anticoagulant SOLUTIONon. It has 15% less citrate ion over citrate-phosphate-dextrose SOLUTIONon with adenine (ACD) and provides improved isotonicity oxygen release function and posttransfusion viability for red blood cells.