Learn More
MOLECULAR DEPOT LLC Citrate Synthase - 200 U
Catalog No. 5000013533
Encompass_Preferred
Citrate Synthase from Porcine Heart consists of a native Citrate Synthase purified from Porcine Heart This enzyme catalyzes the conversion of citrate to acetyl-CoA in the presence of coenzyme-A This product has been used as molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only
Catalog No. 50-000-13533 Supplier MOLECULAR DEPOT LLC Supplier No. B2013107
Citrate Synthase from Porcine Heart consists of a native Citrate Synthase purified from Porcine Heart This enzyme catalyzes the conversion of citrate to acetyl-CoA in the presence of coenzyme-A This product has been used as molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only