Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Citrate synthase | 9027-96-7 | ~98 KDa dimer (49 KDa monomer) | 1000 U
SDP

Catalog No. 5000400365
Encompass_Preferred

Citrate synthase catalyzes the initial reaction of the citric acid cycle, facilitating the condensation of acetyl-CoA and oxaloacetate to produce citrate. This enzyme is primarily found within the mitochondrial matrix of eukaryotic cells.

  • Catalyzes the condensation of acetyl-CoA and oxaloacetate
  • Forms citrate, initiating the citric acid cycle
  • Localized in the mitochondrial matrix of eukaryotic cells
  • Specific activity of ≥100 U/mg protein
  • Unit defined as 1.0 μmole of citrate formed per minute at pH 8.0 at 37°C
  • Liquid appearance
  • Colorless to light yellow
  • Molecular weight of ~98 KDa dimer (49 KDa monomer)

Catalog No. 50-004-00365 Supplier Medchemexpress LLC Supplier No. HYP27391000U
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Citrate synthase catalyzes the initial reaction of the citric acid cycle, facilitating the condensation of acetyl-CoA and oxaloacetate to produce citrate. This enzyme is primarily found within the mitochondrial matrix of eukaryotic cells.

  • Catalyzes the condensation of acetyl-CoA and oxaloacetate
  • Forms citrate, initiating the citric acid cycle
  • Localized in the mitochondrial matrix of eukaryotic cells
  • Specific activity of ≥100 U/mg protein
  • Unit defined as 1.0 μmole of citrate formed per minute at pH 8.0 at 37°C
  • Liquid appearance
  • Colorless to light yellow
  • Molecular weight of ~98 KDa dimer (49 KDa monomer)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.