Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Citrate synthase | 9027-96-7 | MFCD00131495 | ~98 KDa dimer | 250 U
SDP

Catalog No. 5000396212
Encompass_Preferred

Citrate synthase catalyzes the first reaction of the citric acid cycle, condensing acetyl-CoA and oxaloacetate to form citrate. This enzyme is localized within eukaryotic cells in the mitochondrial matrix.

  • Catalyzes the first reaction of the citric acid cycle
  • Condenses acetyl-CoA and oxaloacetate to form citrate
  • Localized in the mitochondrial matrix
  • Specific activity: ≥100 U/mg protein
  • Appearance: Colorless to light yellow liquid
  • Source: Microorganism

Catalog No. 50-003-96212 Supplier Medchemexpress LLC Supplier No. 250UHYP2739
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Citrate synthase catalyzes the first reaction of the citric acid cycle, condensing acetyl-CoA and oxaloacetate to form citrate. This enzyme is localized within eukaryotic cells in the mitochondrial matrix.

  • Catalyzes the first reaction of the citric acid cycle
  • Condenses acetyl-CoA and oxaloacetate to form citrate
  • Localized in the mitochondrial matrix
  • Specific activity: ≥100 U/mg protein
  • Appearance: Colorless to light yellow liquid
  • Source: Microorganism

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.