Learn More
Sigma Aldrich Fine Chemicals Biosciences Citrate Synthase Assay Kit
Catalog No. 502907320
Encompass
The Citrate Synthase Assay Kit contains all the required reagents (including a positive control enzyme) for a fast and simple measurement of citrate synthase activity in a whole cell extract or in isolated mitochondria. In addition the kit enables testing of the intactness of the mitochondrial inner membrane.
Catalog No. 50-290-7320 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CS07201KT
The Citrate Synthase Assay Kit contains all the required reagents (including a positive control enzyme) for a fast and simple measurement of citrate synthase activity in a whole cell extract or in isolated mitochondria. In addition the kit enables testing of the intactness of the mitochondrial inner membrane.