Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Citrate Synthetase Antibody (YA2802) | 10 UL
SDP

Catalog No. 5000444242
Encompass_Preferred

Citrate Synthetase Antibody (YA2802) is a rabbit-derived, non-conjugated IgG monoclonal antibody. It targets Citrate Synthetase and is suitable for various antibody-related applications.

  • Rabbit-derived
  • Non-conjugated IgG monoclonal antibody
  • Targets Citrate Synthetase
  • Recombinant antibody
  • Predicted band size: 52 kDa
  • Observed band size: 45 kDa

Catalog No. 50-004-44242 Supplier Medchemexpress LLC Supplier No. HYP8305710UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Citrate Synthetase Antibody (YA2802) is a rabbit-derived, non-conjugated IgG monoclonal antibody. It targets Citrate Synthetase and is suitable for various antibody-related applications.

  • Rabbit-derived
  • Non-conjugated IgG monoclonal antibody
  • Targets Citrate Synthetase
  • Recombinant antibody
  • Predicted band size: 52 kDa
  • Observed band size: 45 kDa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.