Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Citreoviridin | 25425-12-1 | 1 MG
SDP

Catalog No. 5000419252
Encompass_Preferred

Citreoviridin is a toxin derived from Penicillium citreoviride NRRL 2579. It is known to inhibit brain synaptosomal Na+/K+-ATPase and, in microsomes, significantly stimulate both Na+/K+-ATPase and Mg2+-ATPase activities in a dose-dependent manner. Additionally, Citreoviridin inhibits cell proliferation and enhances apoptosis in human umbilical vein endothelial cells.

  • Toxin from Penicillium citreoviride NRRL 2579
  • Inhibits brain synaptosomal Na+/K+-ATPase
  • Stimulates Na+/K+-ATPase and Mg2+-ATPase in microsomes
  • Inhibits cell proliferation
  • Enhances apoptosis of human umbilical vein endothelial cells
  • Antiproliferative activity against A549 and CL1-0 cells with an IC50 of 1.5 μM
  • Molecular formula: C23H30O6
  • Molecular weight: 402.48
  • Appearance: Light yellow to yellow solid

Catalog No. 50-004-19252 Supplier Medchemexpress LLC Supplier No. HYN67451MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Citreoviridin is a toxin derived from Penicillium citreoviride NRRL 2579. It is known to inhibit brain synaptosomal Na+/K+-ATPase and, in microsomes, significantly stimulate both Na+/K+-ATPase and Mg2+-ATPase activities in a dose-dependent manner. Additionally, Citreoviridin inhibits cell proliferation and enhances apoptosis in human umbilical vein endothelial cells.

  • Toxin from Penicillium citreoviride NRRL 2579
  • Inhibits brain synaptosomal Na+/K+-ATPase
  • Stimulates Na+/K+-ATPase and Mg2+-ATPase in microsomes
  • Inhibits cell proliferation
  • Enhances apoptosis of human umbilical vein endothelial cells
  • Antiproliferative activity against A549 and CL1-0 cells with an IC50 of 1.5 μM
  • Molecular formula: C23H30O6
  • Molecular weight: 402.48
  • Appearance: Light yellow to yellow solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.