Learn More
Medchemexpress LLC HY-N1428 100mg Medchemexpress, Citric acid CAS:77-92-9 Purity:>98%
SDP
Catalog No. 501933831
Encompass_Preferred
Medchemexpress, HY-N1428 100mg Citric acid CAS:77-92-9 Citric acid is a weak organic tricarboxylic acid found in citrus fruits. Citric acid is a natural preservative and food tartness enhancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3831 Supplier Medchemexpress LLC Supplier No. HYN1428100MG
Medchemexpress, HY-N1428 100mg Citric acid CAS:77-92-9 Citric acid is a weak organic tricarboxylic acid found in citrus fruits. Citric acid is a natural preservative and food tartness enhancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.