Learn More
Medchemexpress LLC Citric acid-13C3 | 302912-06-7 | 973254 | 195.10 | 5 MG
SDP
Catalog No. 5000415357
Encompass_Preferred
Citric acid-13C3 is the 13C-labeled Citric acid. It is a natural preservative and food tartness enhancer. Citric acid induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells. Citric acid can cause oxidative damage of the liver by decreasing antioxidative enzyme activities. It is also used as an acidulant, emulsifier, sequestrant, and buffering agent across various industries.
  • 13C-labeled citric acid
  • Natural preservative
  • Food tartness enhancer
  • Induces apoptosis and cell cycle arrest
  • Functions as an acidulant, emulsifier, sequestrant, and buffering agent
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-15357 Supplier Medchemexpress LLC Supplier No. HYN1428S35MG
Citric acid-13C3 is the 13C-labeled Citric acid. It is a natural preservative and food tartness enhancer. Citric acid induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells. Citric acid can cause oxidative damage of the liver by decreasing antioxidative enzyme activities. It is also used as an acidulant, emulsifier, sequestrant, and buffering agent across various industries.
  • 13C-labeled citric acid
  • Natural preservative
  • Food tartness enhancer
  • Induces apoptosis and cell cycle arrest
  • Functions as an acidulant, emulsifier, sequestrant, and buffering agent
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS