Learn More
Medchemexpress LLC Citric acid-13C6 | 287389-42-8 | MFCD01075573 | 99.9% | 198.08 | 13C6H8O7 | 10 MG
SDP
Catalog No. 5000296132
Encompass_Preferred
Isotopically labeled citric acid with all six carbons as 13C, provided as a high-purity stable isotope reagent for use as a tracer or internal standard in analytical methods such as NMR, GC-MS, and LC-MS. Supplied as a 10 mg solid with recommended sealed storage away from moisture.
  • High isotopic enrichment suitable for quantitative analysis.
  • High purity for accurate analytical results.
  • Suitable as tracer or internal standard for NMR, GC-MS, LC-MS.
  • Solid form; easy to weigh and dissolve for standards.
  • Recommended sealed storage away from moisture to preserve stability.
Catalog No. 50-002-96132 Supplier Medchemexpress LLC Supplier No. HYN1428S110MG
Isotopically labeled citric acid with all six carbons as 13C, provided as a high-purity stable isotope reagent for use as a tracer or internal standard in analytical methods such as NMR, GC-MS, and LC-MS. Supplied as a 10 mg solid with recommended sealed storage away from moisture.
  • High isotopic enrichment suitable for quantitative analysis.
  • High purity for accurate analytical results.
  • Suitable as tracer or internal standard for NMR, GC-MS, LC-MS.
  • Solid form; easy to weigh and dissolve for standards.
  • Recommended sealed storage away from moisture to preserve stability.