Learn More
Medchemexpress LLC Citric acid | 2 G
SDP
Catalog No. 5000415323
Encompass_Preferred
Citric acid is a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells. Citric acid can cause oxidative damage of the liver by decreasing antioxidative enzyme activities. It is also an acidulant, emulsifier, sequestrant, and buffering agent widely used across many industries.
  • Natural preservative
  • Food tartness enhancer
  • Induces apoptosis and cell cycle arrest at G2/M and S phases in HaCaT cells
  • Can cause oxidative damage to the liver
  • Functions as an acidulant
  • Functions as an emulsifier
  • Functions as a sequestrant
  • Functions as a buffering agent
Catalog No. 50-004-15323 Supplier Medchemexpress LLC Supplier No. HYN14282G
Citric acid is a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells. Citric acid can cause oxidative damage of the liver by decreasing antioxidative enzyme activities. It is also an acidulant, emulsifier, sequestrant, and buffering agent widely used across many industries.
  • Natural preservative
  • Food tartness enhancer
  • Induces apoptosis and cell cycle arrest at G2/M and S phases in HaCaT cells
  • Can cause oxidative damage to the liver
  • Functions as an acidulant
  • Functions as an emulsifier
  • Functions as a sequestrant
  • Functions as a buffering agent