Learn More
U.S. Pharmacopeia Citric Acid, 77-92-9, MFCD00011669, 200mg
Catalog No. NC9981239
Encompass
Molecular formula C6H8O7, Molecular weight 192.12, Melting Point 305.6 - 318.2 °F (152 - 159 °C), Boiling Point > 347 °F (> 175 °C)
Catalog No. NC9981239 Supplier U.S. Pharmacopeia Supplier No. 1134368
Molecular formula C6H8O7, Molecular weight 192.12, Melting Point 305.6 - 318.2 °F (152 - 159 °C), Boiling Point > 347 °F (> 175 °C)