Learn More
Electron Microscopy Sciences Citric Acid 0.5% Aqueous Solution 500 ML
SDP
Catalog No. 5031686
Encompass_Preferred
Used in Pinkerton's Method for Rickettsia, For inclusion body stain. Aee Simmons, J.S. and Gentzkow, C.J., Laboratory Methods of the United States Army 5th Ed., Lee & Febiger, Philadelphia, p. 572, 1944 or Luna, L.G., (ed) Manual of Histologic Staining Methods or the AFIP, 3rd Ed., McGraw-Hill, N.Y., . 237, c 1968
Catalog No. 50-316-86 Supplier Electron Microscopy Sciences Supplier No. 2613103
Used in Pinkerton's Method for Rickettsia, For inclusion body stain. Aee Simmons, J.S. and Gentzkow, C.J., Laboratory Methods of the United States Army 5th Ed., Lee & Febiger, Philadelphia, p. 572, 1944 or Luna, L.G., (ed) Manual of Histologic Staining Methods or the AFIP, 3rd Ed., McGraw-Hill, N.Y., . 237, c 1968