Learn More
American Radiolabeled Chemicals Inc CITRIC ACID 6-14C 50UC
SDP
Catalog No. 50352922
Encompass_Preferred
Citric acid-6-14C 50uCi in ethanol:water (1:9), 0.1 mCi/ml, 50-60 mCi/mmol, M.W. 192.12, Exclusive
Catalog No. 50-352-922 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 365150UC
Citric acid-6-14C 50uCi in ethanol:water (1:9), 0.1 mCi/ml, 50-60 mCi/mmol, M.W. 192.12, Exclusive