Learn More
Medchemexpress LLC Citric acid | 77-92-9 | MFCD00011669 | 192.12 | 1 ML
SDP
Catalog No. 5000415691
Encompass_Preferred
Citric acid is a natural preservative and food tartness enhancer, widely used as an acidulant, emulsifier, sequestrant, and buffering agent. It can induce apoptosis and cell cycle arrest in HaCaT cells and may cause oxidative damage to the liver by decreasing antioxidative enzyme activities.
  • Induces apoptosis and cell cycle arrest at G2/M and S phases in HaCaT cells.
  • Functions as a natural preservative and food tartness enhancer.
  • Utilized as an acidulant, emulsifier, sequestrant, and buffering agent in various industries.
  • Shows antiproliferative activity in a dose-dependent manner in HaCaT cells.
Catalog No. 50-004-15691 Supplier Medchemexpress LLC Supplier No. HYN142810MM/1ML
Citric acid is a natural preservative and food tartness enhancer, widely used as an acidulant, emulsifier, sequestrant, and buffering agent. It can induce apoptosis and cell cycle arrest in HaCaT cells and may cause oxidative damage to the liver by decreasing antioxidative enzyme activities.
  • Induces apoptosis and cell cycle arrest at G2/M and S phases in HaCaT cells.
  • Functions as a natural preservative and food tartness enhancer.
  • Utilized as an acidulant, emulsifier, sequestrant, and buffering agent in various industries.
  • Shows antiproliferative activity in a dose-dependent manner in HaCaT cells.