Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric acid, 99.91%, MP Biomedicals™

Catalog No. ICN15069905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15069905 5 kg
ICN15069901 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15069905 Supplier MP Biomedicals, Inc Supplier No. 0215069905
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.138
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
PubChem CID 22230
ChEBI CHEBI:31404
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O

Specifications

Color White
Quantity 5 kg
Physical Form Crystalline Powder
Formula Weight 210.1
Percent Purity 99.91%
Chemical Name or Material Citric acid
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.