Learn More
Supply Solutions Avantor Performance Materials - Citric Acid, Anhydrous, Powder. Size: 12Kg; Grade: Usp; Concentration: 99.5 - 100.5%.; Size: 12Kg
Catalog No. 50877605
Encompass_Preferred
Avantor Performance Materials - Citric Acid, Anhydrous, Powder. Size: 12kg; Grade: USP; Concentration: 99.5 - 100.5%.; Size: 12kg; Grade: USP; Concentration: 99.5 - 100.5%; CAS: 77-92-9; Hazardous: N; Container Material: Poly Pail; Formula Weight: 192.12; Chemical Formula: HOC(COOH)(CH?COOH)?; Density: 1.665
This item is not returnable. View return policy
Catalog No. 50-877-605 Supplier Supply Solutions Supplier No. BAKER 012207
Avantor Performance Materials - Citric Acid, Anhydrous, Powder. Size: 12kg; Grade: USP; Concentration: 99.5 - 100.5%.; Size: 12kg; Grade: USP; Concentration: 99.5 - 100.5%; CAS: 77-92-9; Hazardous: N; Container Material: Poly Pail; Formula Weight: 192.12; Chemical Formula: HOC(COOH)(CH?COOH)?; Density: 1.665