Learn More
Research Products International Corp Citric Acid Anhydrous, 500 Grams
SDP
Catalog No. 501368155
Encompass_Preferred
Citric acid anhydrous (C₆H₈O₇) is a white, water-soluble compound (192.12 g/mol) used in life science labs as a buffer and chelating agent. It helps maintain pH in enzymatic reactions, electrophoresis, and cell assays, and binds metal ions to stabilize biomolecules in nucleic acid extraction and protein purification protocols.
Catalog No. 50-136-8155 Supplier Research Products International Corp Supplier No. C34500500.0
Citric acid anhydrous (C₆H₈O₇) is a white, water-soluble compound (192.12 g/mol) used in life science labs as a buffer and chelating agent. It helps maintain pH in enzymatic reactions, electrophoresis, and cell assays, and binds metal ions to stabilize biomolecules in nucleic acid extraction and protein purification protocols.