Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Citric Acid, Anhydrous Reagent A.C.S. 500 GR
SDP

Catalog No. 50980293
Encompass_Preferred

(Piperazine-N,N-bis (2-ethanesulfonic acid,) 1.5 Sodium)
CAS #77-92-9 C₆H₈O₇ F.W. 192.13
Assay: Min. 99.5%

Catalog No. 50-980-293 Supplier Electron Microscopy Sciences Supplier No. 12480
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Piperazine-N,N-bis (2-ethanesulfonic acid,) 1.5 Sodium)
CAS #77-92-9 C₆H₈O₇ F.W. 192.13
Assay: Min. 99.5%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.