Learn More
Medchemexpress LLC Citric acid-d4-1 | 77-92-9 | MFCD00209702 | 97.0% | 196.15 g/mol | C6H4D4O7 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN1428S610MG
Encompass_Preferred
Citric acid-d4-1 is the deuterium-labeled form of citric acid supplied as a research-grade isotopic standard. It contains four deuterium atoms (d4), has formula C6H4D4O7, and is provided as a 10 mg solid for use as an internal standard in mass spectrometry and metabolic tracing.
  • Deuterium-labeled (d4) isotopic standard.
  • Intended for analytical and mass spectrometry applications.
  • Provided as a 10 mg solid for small-batch analytical use.
  • Formula C6H4D4O7 and molecular weight 196.15 g/mol.
  • Manufacturer lists purity of 97.0% on the product page.
Catalog No. 50-002-95717