Learn More
Medchemexpress LLC Citric acid-d4 | 147664-83-3 | 196.15 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN1428S
Encompass_Preferred
Citric acid-d4 is the deuterium labeled Citric acid (HY-N1428). Citric acid is a natural preservative and food tartness enhancer. It induces apoptosis and cell cycle arrest at G2/M phase and S phase in HaCaT cells. Citric acid can cause oxidative damage of the liver by decreasing antioxidative enzyme activities. It is also an acidulant, emulsifier, sequestrant, and buffering agent widely used across many industries. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration has garnered attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89258